siRNA Details
Gene Symbol: Nsun7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017452.1 | NP_001017452.1 | 5, 6 | 953 | ||
XM_006250946.2 | XP_006251008.1 | 5, 6 | 1244 | |||
XM_006250947.2 | XP_006251009.1 | 5, 6 | 1247 | |||
XM_006250948.2 | XP_006251010.1 | 5, 6 | 1244 | |||
GenBank mRNA | BC083653.1 | - | 5, 6 | 953 | 7135159 |
