siRNA Details
Gene Symbol: CT83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017978.3 | NP_001017978.1 | 2 | 412 | ||
GenBank mRNA | AK000618.1 | - | 2 | 382 | ||
AK026566.1 | - | 2 | 382 | |||
BC062223.1 | - | 2 | 359 | 6101827 | ||
HY170569.1 | - | 2 | 413 |
