siRNA Details
Gene Symbol: Bmpr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024259.1 | NP_001019430.1 | 6 | 852 | ||
XM_006233400.2 | XP_006233462.1 | 10 | 1696 | |||
XM_006233401.2 | XP_006233463.1 | 9 | 1357 | |||
XM_008761513.1 | XP_008759735.1 | 9 | 1556 | |||
XM_008761514.1 | XP_008759736.1 | 12 | 2078 | |||
GenBank mRNA | BC092609.1 | - | 6 | 852 | 7310757 |
