siRNA Details
Gene Symbol: ZNF334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270497.1 | NP_001257426.1 | 5 | 472 | ||
NM_018102.4 | NP_060572.3 | 4 | 412 | |||
NR_073026.1 | 4 | 380 | ||||
XM_011528892.2 | XP_011527194.1 | 5 | 1917 | |||
XM_011528895.2 | XP_011527197.1 | 4 | 1407 | |||
XM_011528897.2 | XP_011527199.1 | 4 | 1712 | |||
XM_017027933.1 | XP_016883422.1 | 3 | 1525 | |||
XM_017027934.1 | XP_016883423.1 | 6 | 675 | |||
XM_017027935.1 | XP_016883424.1 | 5 | 1323 | |||
XM_017027936.1 | XP_016883425.1 | 4 | 1281 | |||
XM_017027937.1 | XP_016883426.1 | 3 | 1637 | |||
XM_017027938.1 | XP_016883427.1 | 4 | 325 | |||
XM_017027939.1 | XP_016883428.1 | 5 | 1425 | |||
XM_017027940.1 | XP_016883429.1 | 5 | 559 | |||
XM_017027941.1 | XP_016883430.1 | 4 | 1344 | |||
XM_017027942.1 | XP_016883431.1 | 3 | 1578 | |||
XM_017027943.1 | XP_016883432.1 | 3 | 1276 | |||
XM_017027944.1 | XP_016883433.1 | 2 | 1460 | |||
GenBank mRNA | AK001331.1 | - | 4 | 407 | ||
AK057717.1 | - | 3 | 702 | |||
AK095611.1 | - | 5 | 468 | |||
AL833936.1 | - | 3 | 589 | |||
BC024177.2 | - | 4 | 408 | 4824096 | ||
BC026321.1 | - | 5 | 1434 | 4819292 | ||
BG720863.1 | - | 4 | 413 | |||
DA080528.1 | - | 4 | 348 |
