siRNA Details
Gene Symbol: THAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018105.2 | NP_060575.1 | 2 | 356 | ||
GenBank mRNA | AK001339.1 | - | 2 | 350 | ||
AK223231.1 | - | 2 | 335 | |||
BC021721.2 | - | 2 | 330 | 4795229 | ||
BG497522.1 | - | 2 | 357 | |||
CD709111.1 | - | 2 | 366 | |||
CR457256.1 | - | 2 | 119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
