siRNA Details
Gene Symbol: Lrrc8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024782.1 | NP_001019953.1 | 4 | 2908 | ||
XM_006233814.2 | XP_006233876.1 | 2 | 2754 | |||
XM_008761660.1 | XP_008759882.1 | 2 | 2754 | |||
GenBank mRNA | BC097371.1 | - | 4 | 2908 | 7444090 |
