siRNA Details
Gene Symbol: Osgepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024787.1 | NP_001019958.1 | 4 | 855 | ||
XM_006244893.2 | XP_006244955.1 | 4 | 904 | |||
XM_006244894.2 | XP_006244956.1 | 4 | 868 | |||
GenBank mRNA | BC078974.1 | - | 4 | 875 | ||
BC097950.1 | - | 4 | 855 | 7389045 |
