siRNA Details
Gene Symbol: Usp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024790.1 | NP_001019961.1 | 9, 10 | 987 | ||
XM_006245756.2 | XP_006245818.1 | 10, 11 | 1199 | |||
XM_006245757.2 | XP_006245819.1 | 9, 10 | 2036 | |||
XM_006245758.2 | XP_006245820.1 | 9, 10 | 1129 | |||
XM_006245759.1 | XP_006245821.1 | 9, 10 | 1080 | |||
GenBank mRNA | AY641530.1 | - | 9, 10 | 987 |
