siRNA Details
Gene Symbol: Pomk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024883.1 | NP_001020054.1 | 5 | 2717 | ||
XM_008771353.1 | XP_008769575.1 | 3 | 2491 | |||
GenBank mRNA | BC097466.1 | - | 5 | 2717 | 7457667 | |
FQ211927.1 | - | 5 | 2707 |
