siRNA Details
Gene Symbol: RGD1306186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024986.1 | NP_001020157.1 | 3 | 1353 | ||
GenBank mRNA | BC090317.1 | - | 3 | 1353 | 7305376 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1306186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024986.1 | NP_001020157.1 | 3 | 1353 | ||
GenBank mRNA | BC090317.1 | - | 3 | 1353 | 7305376 |