siRNA Details
Gene Symbol: LOC308990 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025001.1 | NP_001020172.1 | 2 | 1483 | ||
XM_006230265.2 | XP_006230327.1 | 2 | 1286 | |||
XM_006230266.2 | XP_006230328.1 | 2 | 1427 | |||
XM_006230267.2 | XP_006230329.1 | 2 | 1385 | |||
XM_006230268.2 | XP_006230330.1 | 2 | 1244 | |||
GenBank mRNA | BC091205.1 | - | 1 | 1266 | 7380611 | |
BC091216.1 | - | 2 | 1483 | 7383904 |
