siRNA Details
Gene Symbol: Ifi202b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF140672.1 | - | 6 | 1394 | ||
BC018233.1 | - | 1520 | 3990226 | |||
BC055888.1 | - | 5 | 1513 | 4945964 | ||
DQ222946.1 | - | 5 | 1300 | |||
JX945582.1 | - | 5 | 1546 | |||
M31418.1 | - | 1593 |
