siRNA Details
Gene Symbol: Fat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081286.2 | NP_001074755.2 | 1 | 2270 | ||
GenBank mRNA | AJ250768.1 | - | 1 | 2270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
