siRNA Details
Gene Symbol: Myh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010855.3 | NP_034985.2 | 4 | 330 | ||
GenBank mRNA | AK138608.1 | - | 4 | 331 | ||
AY963801.1 | - | 4 | 330 | |||
BC141362.1 | - | 2 | 247 | 9055974 | ||
BG796256.1 | - | 3 | 351 | |||
DQ066414.1 | - | 421 | ||||
DQ066415.1 | - | 4 | 515 | |||
DQ066416.1 | - | 4 | 417 | |||
DQ066417.1 | - | 4 | 372 | |||
DQ066418.1 | - | 4 | 377 | |||
DQ066419.1 | - | 4 | 448 | |||
DQ066420.1 | - | 2 | 286 | |||
DQ233297.1 | - | 3 | 283 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
