siRNA Details
Gene Symbol: KIAA0895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100425.1 | NP_001093895.1 | 3 | 538 | ||
NM_001199706.1 | NP_001186635.1 | 2 | 627 | |||
NM_001199707.1 | NP_001186636.1 | 2 | 779 | |||
NM_001199708.1 | NP_001186637.1 | 2 | 468 | |||
NM_015314.2 | NP_056129.2 | 2 | 635 | |||
XM_005249689.3 | XP_005249746.1 | 2 | 844 | |||
GenBank mRNA | AB020702.1 | - | 2 | 549 | ||
AK293912.1 | - | 2 | 400 | |||
AK294686.1 | - | 2 | 595 | |||
AK302559.1 | - | 2 | 595 | |||
BC028678.2 | - | 3 | 500 | 4836984 | ||
BC056191.1 | - | 2 | 519 | 5298453 | ||
BC110422.1 | - | 2 | 779 | 5199115 | ||
BC136362.1 | - | 2 | 529 | 9020349 | ||
BC144032.1 | - | 2 | 532 | 9052552 | ||
DC403070.1 | - | 2 | 468 |
