siRNA Details
Gene Symbol: KIAA0895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100425.1 | NP_001093895.1 | 7 | 3414 | ||
NM_001199706.1 | NP_001186635.1 | 7 | 3647 | |||
NM_001199707.1 | NP_001186636.1 | 6 | 3655 | |||
NM_001199708.1 | NP_001186637.1 | 7 | 3488 | |||
NM_001300956.1 | NP_001287885.1 | 6 | 2799 | |||
NM_015314.2 | NP_056129.2 | 6 | 3511 | |||
XM_005249689.3 | XP_005249746.1 | 7 | 3864 | |||
XM_005249690.4 | XP_005249747.1 | 6 | 2961 | |||
GenBank mRNA | AB020702.1 | - | 6 | 3425 | ||
BC028678.2 | - | 7 | 3371 | 4836984 | ||
BC056191.1 | - | 6 | 3395 | 5298453 | ||
BC136362.1 | - | 6 | 3405 | 9020349 | ||
BC144032.1 | - | 7 | 3552 | 9052552 |
