siRNA Details
Gene Symbol: CNR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160226.1 | NP_001153698.1 | 3 | 2850 | ||
NM_001160258.1 | NP_001153730.1 | 4 | 2888 | |||
NM_001160259.1 | NP_001153731.1 | 2 | 2763 | |||
NM_016083.4 | NP_057167.2 | 2 | 2707 | |||
NM_033181.3 | NP_149421.2 | 2 | 2362 | |||
XM_006715330.3 | XP_006715393.1 | 2 | 2654 | |||
XM_011535424.1 | XP_011533726.1 | 3 | 3194 | |||
XM_011535425.1 | XP_011533727.1 | 4 | 3232 | |||
XM_011535426.1 | XP_011533728.1 | 5 | 3390 | |||
XM_011535428.1 | XP_011533730.1 | 3 | 3041 | |||
XM_017010238.1 | XP_016865727.1 | 4 | 6018 | |||
XM_017010239.1 | XP_016865728.1 | 3 | 4341 | |||
XM_017010240.1 | XP_016865729.1 | 2 | 2701 | |||
GenBank mRNA | BX538191.1 | - | 1 | 71 | ||
DA128033.1 | - | 1 | 525 | |||
DA239984.1 | - | 1 | 547 | |||
DA793373.1 | - | 1 | 36 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
