siRNA Details
Gene Symbol: Draxin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027426.3 | NP_081702.2 | 7 | 2494 | ||
GenBank mRNA | AK086198.1 | - | 7 | 2495 | ||
BC057066.1 | - | 1 | 364 | |||
BC058734.1 | - | 3 | 1610 | |||
BC060239.1 | - | 7 | 2475 | 6853328 |
