siRNA Details
Gene Symbol: 9130019O22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030226.3 | NP_084502.2 | 3 | 1996 | ||
GenBank mRNA | AK020267.1 | - | 3 | 1871 | ||
AK035847.1 | - | 3 | 1998 | |||
BC031466.1 | - | 3 | 1966 | 4038184 |
