siRNA Details
Gene Symbol: Celsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191110.1 | NP_001178039.1 | 1 | 1593 | ||
XM_006233192.2 | XP_006233254.1 | 1 | 2123 | |||
GenBank mRNA | AY212289.1 | - | 1 | 176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
