siRNA Details
Gene Symbol: Celsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191110.1 | NP_001178039.1 | 12, 13 | 5611 | ||
XM_006233192.2 | XP_006233254.1 | 12, 13 | 6141 | |||
GenBank mRNA | AB011529.1 | - | 12, 13 | 3286 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
