siRNA Details
Gene Symbol: Ppp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134359.1 | NP_599186.1 | 7 | 903 | ||
XM_006230206.1 | XP_006230268.1 | 8 | 899 | |||
GenBank mRNA | BC091574.1 | - | 7 | 903 | 7135215 | |
FQ216602.1 | - | 8 | 859 | |||
M58443.1 | - | 1 | 122 |
