siRNA Details
Gene Symbol: 9530068E07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153117.2 | NP_694757.1 | 3 | 1027 | ||
GenBank mRNA | AK079269.1 | - | 3 | 1028 | ||
AK135934.1 | - | 3 | 991 | |||
AK144677.1 | - | 3 | 992 | |||
AK145254.1 | - | 3 | 997 | |||
AK147956.1 | - | 3 | 994 | |||
AK150517.1 | - | 3 | 1011 | |||
AK154955.1 | - | 3 | 997 | |||
BC006957.1 | - | 3 | 881 | |||
BC034829.1 | - | 3 | 1001 | 4482756 |
