siRNA Details
Gene Symbol: A530064D06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113556.1 | NP_001107028.1 | 3 | 1476 | ||
NM_178796.6 | NP_848911.2 | 4 | 1660 | |||
GenBank mRNA | AK080114.1 | - | 4 | 1621 | ||
AK089248.1 | - | 3 | 1490 | |||
AK171433.1 | - | 1437 |
