siRNA Details
Gene Symbol: ZNF600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321866.1 | NP_001308795.1 | 6 | 3680 | ||
NM_001321867.1 | NP_001308796.1 | 4 | 3057 | |||
NM_198457.3 | NP_940859.3 | 3 | 2930 | |||
GenBank mRNA | BX640933.1 | - | 3 | 2902 |
