siRNA Details
Gene Symbol: Slc16a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198760.1 | NP_942055.1 | 6 | 1916 | ||
XM_006247613.2 | XP_006247675.1 | 7 | 4594 | |||
XM_006247614.2 | XP_006247676.1 | 7 | 2615 | |||
XM_006247617.2 | XP_006247679.1 | 8 | 2299 | |||
XM_006247618.2 | XP_006247680.1 | 7 | 2181 | |||
XM_006247619.2 | XP_006247681.1 | 6 | 4137 | |||
XM_006247620.2 | XP_006247682.1 | 6 | 3818 | |||
XM_008768375.1 | XP_008766597.1 | 6 | 2427 | |||
XM_008768376.1 | XP_008766598.1 | 7 | 4325 | |||
GenBank mRNA | AY325107.1 | - | 6 | 1916 | ||
BC081999.1 | - | 6 | 1977 | 7131044 | ||
FQ212616.1 | - | 2 | 1252 |
