siRNA Details
Gene Symbol: Srsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009720.2 | NP_001009720.1 | 2 | 819 | ||
XM_006247813.2 | XP_006247875.1 | 2 | 846 | |||
XM_008768382.1 | XP_008766604.1 | 2 | 846 | |||
GenBank mRNA | BC058508.1 | - | 2 | 823 | 6889746 | |
FQ232539.1 | - | 2 | 821 |
