siRNA Details
Gene Symbol: FLJ25758 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024372.1 | 1 | 543 | |||
GenBank mRNA | AK098624.1 | - | 1 | 550 | ||
AY090781.1 | - | 1 | 543 |
Gene Symbol: LOC100034248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128820.1 | - | 1 | 548 |
