siRNA Details
Gene Symbol: Ccdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011561.1 | NP_001011561.1 | 3 | 484 | ||
GenBank mRNA | BC079021.1 | - | 3 | 484 | 7113858 |
Gene Symbol: LOC679727 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751971.3 | XP_003752019.2 | 1 | 182 |
