siRNA Details
Gene Symbol: Ccdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011561.1 | NP_001011561.1 | 6 | 918 | ||
GenBank mRNA | BC079021.1 | - | 6 | 918 | 7113858 |

More Information
Other Species Targeted

