siRNA Details
Gene Symbol: LOC497938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017475.1 | NP_001017475.1 | 3 | 383 | ||
XM_008767853.1 | XP_008766075.1 | 2 | 412 | |||
XM_008767854.1 | XP_008766076.1 | 2 | 412 | |||
GenBank mRNA | BC079298.1 | - | 3 | 383 | 7134515 |
