siRNA Details
Gene Symbol: Tex19.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017482.1 | NP_001017482.1 | 3 | 519 | ||
GenBank mRNA | BC083916.1 | - | 3 | 519 | 7134696 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tex19.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017482.1 | NP_001017482.1 | 3 | 519 | ||
GenBank mRNA | BC083916.1 | - | 3 | 519 | 7134696 |