siRNA Details
Gene Symbol: LOC498750 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017510.3 | NP_001017510.1 | 1 | 447 | ||
GenBank mRNA | AY325226.1 | - | 1 | 447 | ||
BC085940.1 | - | 3 | 908 | 7123624 | ||
BC089212.1 | - | 3 | 706 | 7302835 |
Gene Symbol: LOC100911664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751691.3 | XP_003751739.1 | 3 | 2623 |
