siRNA Details
Gene Symbol: Tex36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024288.1 | NP_001019459.1 | 186 | |||
GenBank mRNA | BC079265.1 | - | 186 | 7133623 |
Gene Symbol: LOC103691249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759961.1 | XP_008758183.1 | 2 | 242 |
