siRNA Details
Gene Symbol: LOC498154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025033.1 | NP_001020204.1 | 5 | 759 | ||
XM_006248959.2 | XP_006249021.1 | 4 | 1242 | |||
GenBank mRNA | BC088429.1 | - | 5 | 759 | 7124272 |
