siRNA Details
Gene Symbol: Fam107b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025034.1 | NP_001020205.1 | 4 | 2135 | ||
XM_008771879.1 | XP_008770101.1 | 4 | 2535 | |||
XM_008771880.1 | XP_008770102.1 | 4 | 2158 | |||
XM_008771881.1 | XP_008770103.1 | 4 | 2584 | |||
XM_008771882.1 | XP_008770104.1 | 4 | 2171 | |||
XM_008771883.1 | XP_008770105.1 | 4 | 2788 | |||
XM_008771884.1 | XP_008770106.1 | 4 | 2180 | |||
GenBank mRNA | BC085116.1 | - | 4 | 2135 | 7107468 |
