siRNA Details
Gene Symbol: Tmem192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163747.1 | NP_001157219.1 | 3 | 333 | ||
NM_028427.3 | NP_082703.1 | 3 | 333 | |||
GenBank mRNA | AK013998.1 | - | 3 | 332 | ||
AK046181.1 | - | 3 | 305 | |||
AK132619.1 | - | 3 | 300 | |||
AK168146.1 | - | 3 | 316 | |||
AK215398.1 | - | 2 | 147 | |||
BC020157.1 | - | 3 | 264 | 3708193 |
