siRNA Details
Gene Symbol: Samd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037221.2 | NP_001032298.1 | 13 | 3499 | ||
NM_001163433.1 | NP_001156905.1 | 12 | 2979 | |||
NM_028966.3 | NP_083242.1 | 12 | 3235 | |||
GenBank mRNA | AK034323.1 | - | 11 | 2829 | ||
AK133470.1 | - | 12 | 3217 | |||
AK135279.1 | - | 1 | 211 | |||
DQ278487.1 | - | 12 | 3012 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
