siRNA Details
Gene Symbol: HEATR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018072.5 | NP_060542.4 | 31, 32 | 4585 | ||
GenBank mRNA | AK098212.1 | - | 6, 7 | 856 | ||
AK125663.1 | - | 13, 14 | 1802 | |||
AK304129.1 | - | 11, 12 | 1662 | |||
AK316413.1 | - | 11, 12 | 1556 | |||
BC035302.1 | - | 2, 3 | 160 | |||
BC065205.1 | - | 9, 10 | 1321 | |||
BC130000.1 | - | 16, 17 | 2509 | |||
BC150614.1 | - | 30, 31 | 4455 | 9056984 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
