siRNA Details
Gene Symbol: TAS2R43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC101155.1 | - | 3 | 574 | 40015679 |
Gene Symbol: TAS2R45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176886.2 | NP_795367.2 | 3 | 574 | ||
GenBank mRNA | AF264628.1 | - | 1 | 45 | ||
BC101154.1 | - | 3 | 574 | 40015678 | ||
BC101156.1 | - | 3 | 574 | 40015680 |
