siRNA Details
Gene Symbol: AURKC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015878.1 | NP_001015878.1 | 6, 7 | 945 | ||
NM_001015879.1 | NP_001015879.1 | 6, 7 | 804 | |||
NM_003160.2 | NP_003151.2 | 6, 7 | 809 | |||
GenBank mRNA | AB017332.1 | - | 6, 7 | 950 | ||
AF054621.1 | - | 6, 7 | 950 | |||
AF059681.1 | - | 6, 7 | 1005 | |||
AY661554.1 | - | 6, 7 | 804 | |||
AY714054.1 | - | 6, 7 | 747 | |||
BC075064.2 | - | 5, 6 | 698 | 30915373 | ||
BX092186.1 | - | 1, 2 | 83 | |||
EU668345.1 | - | 6, 7 | 1005 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
