siRNA Details
Gene Symbol: Olr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000117.1 | NP_001000117.1 | 1 | 740 |
Gene Symbol: LOC100910993 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752171.3 | XP_003752219.1 | 1 | 809 |
