siRNA Details
Gene Symbol: TMEM38B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018112.2 | NP_060582.1 | 6 | 1626 | ||
XM_005252075.2 | XP_005252132.1 | 6 | 2073 | |||
XM_005252076.2 | XP_005252133.1 | 7 | 1643 | |||
XM_005252077.3 | XP_005252134.1 | 7 | 1646 | |||
XM_011518829.1 | XP_011517131.1 | 7 | 1635 | |||
XM_011518832.2 | XP_011517134.1 | 4 | 1346 | |||
XM_011518833.2 | XP_011517135.1 | 7 | 1758 | |||
GenBank mRNA | AK001355.1 | - | 6 | 1607 |
