siRNA Details
Gene Symbol: LMBR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300750.1 | NP_001287679.1 | 18 | 2537 | ||
NM_001300751.1 | NP_001287680.1 | 17 | 2275 | |||
NM_018113.3 | NP_060583.2 | 17 | 2335 | |||
XM_011538563.2 | XP_011536865.1 | 17 | 2489 | |||
XM_011538564.2 | XP_011536866.1 | 17 | 2862 | |||
XM_011538565.2 | XP_011536867.1 | 16 | 3111 | |||
XM_011538566.2 | XP_011536868.1 | 16 | 2085 | |||
XM_017019626.1 | XP_016875115.1 | 17 | 2737 | |||
XM_017019627.1 | XP_016875116.1 | 18 | 2466 | |||
XM_017019628.1 | XP_016875117.1 | 16 | 2162 | |||
XM_017019629.1 | XP_016875118.1 | 18 | 3321 | |||
XM_017019630.1 | XP_016875119.1 | 14 | 4297 | |||
XM_017019631.1 | XP_016875120.1 | 16 | 2025 | |||
GenBank mRNA | AK001356.1 | - | 18 | 2318 | ||
AK001651.1 | - | 17 | 2225 | |||
AK093570.1 | - | 11 | 2068 | |||
BC015015.1 | - | 17 | 2064 | 3910520 | ||
BC031550.1 | - | 17 | 2096 | 5194023 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
