siRNA Details
Gene Symbol: H2-DMb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010388.4 | NP_034518.1 | 776 | |||
GenBank mRNA | AK154923.1 | - | 5, 6 | 880 | ||
BC052864.1 | - | 6, 7 | 1031 | |||
BC141105.1 | - | 5, 6 | 842 | 9055717 | ||
BC145385.1 | - | 5, 6 | 842 | 9053943 | ||
BC145386.1 | - | 5, 6 | 842 | 9053944 | ||
BC171960.1 | - | 5, 6 | 842 | 9054246 | ||
KJ650224.1 | - | 776 | ||||
U35334.1 | - | 776 | ||||
U35336.1 | - | 776 | ||||
U35338.1 | - | 776 |
