siRNA Details
Gene Symbol: Xlr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110784.1 | NP_001104254.1 | 9 | 1509 | ||
GenBank mRNA | BC003812.1 | - | 9 | 1455 | 3601144 | |
BC093495.1 | - | 1432 | 3593889 | |||
CJ302076.1 | - | 1 | 217 | |||
U02599.1 | - | 8 | 1402 |
Gene Symbol: Xlr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK146398.1 | - | 8 | 2248 |
Gene Symbol: Xlr3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | L22977.1 | - | 1434 |
