siRNA Details
Gene Symbol: Rpl34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287581.1 | NP_001274510.1 | 1 | 63 | ||
NM_026724.2 | NP_081000.1 | 1 | 63 | |||
GenBank mRNA | BC058118.1 | - | 1 | 34 | 5683990 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
