siRNA Details
Gene Symbol: Pde4dip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039376.2 | NP_001034465.2 | 47 | 7948 | ||
NM_001289701.1 | NP_001276630.1 | 47 | 7999 | |||
NM_001289702.1 | NP_001276631.1 | 46 | 7846 | |||
GenBank mRNA | AJ290946.1 | - | 1 | 322 | ||
AK040750.1 | - | 8 | 1586 | |||
BC022211.1 | - | 1 | 430 | |||
BC046620.1 | - | 1 | 505 | |||
BC141185.1 | - | 47 | 7999 | 9055797 | ||
BC145504.1 | - | 46 | 7846 | 9054067 |
