siRNA Details
Gene Symbol: A830039N20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK043847.1 | - | 1 | 1256 | ||
AK045914.1 | - | 1 | 1112 | |||
BB269387.2 | - | 1 | 643 | |||
BC038501.1 | - | 1 | 498 |

More Information
Other Species Targeted

