siRNA Details
Gene Symbol: UBE2V1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257393.1 | NP_001244322.1 | 1 | 114 | ||
NM_001257394.1 | NP_001244323.1 | 1 | 114 | |||
NM_001282575.2 | NP_001269504.1 | 1 | 114 | |||
NM_021988.5 | NP_068823.2 | 1 | 114 | |||
NM_022442.5 | NP_071887.1 | 1 | 114 | |||
NM_199144.2 | NP_954595.1 | 1 | 114 | |||
GenBank mRNA | AK314754.1 | - | 1 | 102 | ||
BC005215.1 | - | 1 | 112 | |||
BU540046.1 | - | 1 | 115 | |||
CA453411.1 | - | 112 | ||||
CD103154.1 | - | 1 | 43 | |||
CN346855.1 | - | 1 | 46 | |||
DA409547.1 | - | 1 | 114 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
